APO466131526 Apollo Scientific Ltd

Methyl 3-fluoro-4-methoxybenzoate

  • MDL number MFCD00215839
  • Cas Number : 369-30-2
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 69-73
Iupac Name methyl 3-fluoro-4-methoxybenzoate
Inchi Code 1S/C9H9FO3/c1-12-8-4-3-6(5-7(8)10)9(11)13-2/h3-5H,1-2H3
Inchi Key MIKOKYAREMINFF-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 69-73
Product Link https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-methoxybenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-methoxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart