APO466131527 Apollo Scientific Ltd

4-Fluoro-L-phenylalanine

  • MDL number MFCD00063064
  • Cas Number : 1132-68-9
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 247-249
Iupac Name (2R)-2-amino-3-(4-fluorophenyl)propanoic acid
Inchi Code 1S/C9H10FNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m0/s1
Inchi Key XWHHYOYVRVGJJY-QMMMGPOBSA-N

Description

Synonym (2S)-2-Amino-3-(4-fluorophenyl)propanoic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 247-249
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-l-phenylalanine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-l-phenylalanine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart