APO491316297 Apollo Scientific Ltd

6-Methoxypyridine-2-boronic acid

  • MDL number MFCD03093332
  • Cas Number : 372963-51-4
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-methoxypyridine-2-boronic-acid

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/6-methoxypyridine-2-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart