APO491316370 Apollo Scientific Ltd

6-(4-Morpholinyl)-3-pyridinylboronic acid

  • MDL number MFCD03086116
  • Cas Number : 904326-93-8
  • Molecular Weight : 208.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-4-morpholinyl-3-pyridinylboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-4-morpholinyl-3-pyridinylboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart