APO491316808 Apollo Scientific Ltd

Boric acid, pinacol ester

  • MDL number MFCD15143603
  • Cas Number : 25240-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=79171

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=79171
pictograms GHS07;GHS08
hstatements H302;H315;H319;H360FD
Availability details for
Quantity
  • Availability
  • Add to Cart