APO491317436 Apollo Scientific Ltd

3-Methoxynaphthalene-2-boronic acid

  • MDL number MFCD12025968
  • Cas Number : 104115-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-methoxynaphthalene-2-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxynaphthalene-2-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart