APO491317993 Apollo Scientific Ltd

5-Fluoro-2-(methoxycarbonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD16996304
  • Cas Number : 1400976-17-1
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Inchi Code 1S/C14H18BFO4/c1-13(2)14(3,4)20-15(19-13)11-8-9(16)6-7-10(11)12(17)18-5/h6-8H,1-5H3
Inchi Key RVMVLLXHYRJPOK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxycarbonylphenylboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxycarbonylphenylboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart