APO491318077 Apollo Scientific Ltd

2-Bromo-4,6-difluorophenylboronic acid

  • MDL number MFCD09751350
  • Cas Number : 1315339-48-0
  • Molecular Weight : 236.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4,6-difluorophenylboronic acid
Inchi Code 1S/C6H4BBrF2O2/c8-4-1-3(9)2-5(10)6(4)7(11)12/h1-2,11-12H
Inchi Key POXLFROUJDNFPQ-UHFFFAOYSA-N

Description

Synonym (2-Bromo-4,6-difluorophenyl)boronic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-46-difluorophenylboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-46-difluorophenylboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart