APO491440299 Apollo Scientific Ltd

3,4'-Biphthalic anhydride

  • MDL number MFCD09030654
  • Cas Number : 36978-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-205
Synonym Biphenyl-2,3,3',4'-tetracarboxylic acid dianhydride, 4,5'-Bi-2-benzofuran-1,1',3,3'-tetrone
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48943

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48943
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart