APO491440323 Apollo Scientific Ltd

2,6-Difluoro-3,5-dinitrotoluene

  • MDL number MFCD16620286
  • Cas Number : 112822-76-1
  • Molecular Weight : 218.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Difluoro-3-methyl-1,5-dinitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-35-dinitrotoluene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-35-dinitrotoluene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart