APO491537462 Apollo Scientific Ltd

4-(Ethylsulphonyl)benzaldehyde

  • MDL number MFCD09264545
  • Cas Number : 50899-03-1
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-ethylsulphonylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-ethylsulphonylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart