APO491537504 Apollo Scientific Ltd

(3R)-Morpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD06809588
  • Cas Number : 1187928-88-6
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3r-morpholine-3-carboxylic-acid-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3r-morpholine-3-carboxylic-acid-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart