APO491537550 Apollo Scientific Ltd

3-Bromo-5-(trifluoromethyl)-1H-indazole

  • MDL number MFCD09998163
  • Cas Number : 1086378-32-6
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-(trifluoromethyl)-1H-indazole
Inchi Code 1S/C8H4BrF3N2/c9-7-5-3-4(8(10,11)12)1-2-6(5)13-14-7/h1-3H,(H,13,14)
Inchi Key NJWMXXMBNBUURG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-trifluoromethyl-1h-indazole

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-trifluoromethyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart