APO491537581 Apollo Scientific Ltd

4-(Trifluoromethyl)-1H-indazole

  • MDL number MFCD09263211
  • Cas Number : 1000339-98-9
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-109
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethyl-1h-indazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart