APO491607057 Apollo Scientific Ltd

3,3-Diethoxy-1-propenylboronic acid, pinacol ester

  • MDL number MFCD03788737
  • Cas Number : 153737-25-8
  • Molecular Weight : 256.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/33-diethoxy-1-propenylboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/33-diethoxy-1-propenylboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart