APO497443790 Apollo Scientific Ltd

3,4-Dichloro-D-phenylalanine

  • MDL number MFCD00671682
  • Cas Number : 52794-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2R)-2-Amino-3-(3,4-dichlorophenyl)propanoic acid
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40341

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40341
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart