APO497443799 Apollo Scientific Ltd

5-Hydroxypyrimidine

  • MDL number MFCD06412562
  • Cas Number : 26456-59-7
  • Molecular Weight : 96.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 216(dec.)
Iupac Name 5(2H)-pyrimidinone
Inchi Code 1S/C4H4N2O/c7-4-1-5-3-6-2-4/h1-3,7H
Inchi Key LTXJLQINBYSQFU-UHFFFAOYSA-N

Description

Synonym Pyrimidin-5-ol, 5-Hydroxy-1,3-diazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 216(dec.)
Product Link https://store.apolloscientific.co.uk/product/5-hydroxypyrimidine

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxypyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart