APO497443809 Apollo Scientific Ltd

4'-(Cyanomethyl)acetanilide

  • MDL number MFCD00016398
  • Cas Number : 25025-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Synonym 4-Acetamidophenylacetonitrile, N-[4-(Cyanomethyl)phenyl]acetamide
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12350

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12350
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart