APO497443823 Apollo Scientific Ltd

(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol

  • MDL number MFCD18432555
  • Cas Number : 17288-47-0
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 198-200
Iupac Name 1H-pyrrolo[3,2-b]pyridin-2-ylmethanol
Inchi Code 1S/C8H8N2O/c11-5-6-4-8-7(10-6)2-1-3-9-8/h1-4,10-11H,5H2
Inchi Key NASWAERXWIHATE-UHFFFAOYSA-N

Description

Synonym 2-(Hydroxymethyl)-1H-pyrrolo[3,2-b]pyridine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 198-200
Product Link https://store.apolloscientific.co.uk/product/1h-pyrrolo32-bpyridin-2-ylmethanol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1h-pyrrolo32-bpyridin-2-ylmethanol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart