APO497443861 Apollo Scientific Ltd

Benzyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD09608058
  • Cas Number : 31865-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cbz-6-oxa-3-azabicyclo[3.1.0]hexane
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=80859

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=80859
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart