APO497443867 Apollo Scientific Ltd

D-Alanine isopropyl ester hydrochloride

  • MDL number MFCD17976853
  • Cas Number : 39613-92-8
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 85
Iupac Name isopropyl (2R)-2-aminopropanoate hydrochloride
Inchi Code 1S/C6H13NO2.ClH/c1-4(2)9-6(8)5(3)7;/h4-5H,7H2,1-3H3;1H/t5-;/m1./s1
Inchi Key YAQKNCSWDMGPOY-NUBCRITNSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85
Product Link https://store.apolloscientific.co.uk/product/d-alanine-isopropyl-ester-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/d-alanine-isopropyl-ester-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart