APO497443903 Apollo Scientific Ltd

2-Chloro-5-fluorophenylhydrazine hydrochloride

  • MDL number MFCD03094167
  • Cas Number : 502496-25-5
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name 1-(2-chloro-5-fluorophenyl)hydrazine hydrochloride
Inchi Code 1S/C6H6ClFN2.ClH/c7-5-2-1-4(8)3-6(5)10-9;/h1-3,10H,9H2;1H
Inchi Key DDHOUQFXUDHPDU-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 1-Chloro-4-fluoro-2-hydrazinobenzene hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluorophenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluorophenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart