APO497443907 Apollo Scientific Ltd

3-Chloro-4-fluoroanisole

  • MDL number MFCD00070778
  • Cas Number : 202925-07-3
  • Molecular Weight : 160.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.26
Flash Point 79
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 189
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluoroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluoroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart