APO497443908 Apollo Scientific Ltd

1,1,1-Trifluorohexane-2,4-dione

  • MDL number MFCD00051306
  • Cas Number : 400-54-4
  • Molecular Weight : 168.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Trifluoroacetyl)butan-2-one, 2,4-Dioxo-1,1,1-trifluorohexane
Purity 95
Flash Point 28
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 125
Product Link https://store.apolloscientific.co.uk/product/111-trifluorohexane-24-dione

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/111-trifluorohexane-24-dione
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart