APO497443933 Apollo Scientific Ltd

1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene

  • MDL number MFCD06657548
  • Cas Number : 168971-68-4
  • Molecular Weight : 259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
Inchi Code 1S/C7H3BrF4O/c8-5-2-1-4(3-6(5)9)13-7(10,11)12/h1-3H
Inchi Key KZMHSGBETSENAT-UHFFFAOYSA-N

Description

Synonym 4-Bromo-3-fluorophenyl trifluoromethyl ether
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-2-fluoro-4-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-2-fluoro-4-trifluoromethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart