APO497443939 Apollo Scientific Ltd

5-Bromo-2-chloro-4-(trifluoromethyl)pyrimidine

  • MDL number MFCD13176600
  • Cas Number : 785777-92-6
  • Molecular Weight : 261.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp -6 to -5
Iupac Name 5-bromo-2-chloro-4-(trifluoromethyl)pyrimidine
density 1.879
Inchi Code 1S/C5HBrClF3N2/c6-2-1-11-4(7)12-3(2)5(8,9)10/h1H
Inchi Key VTIPESROQQNDJL-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-chloro-4-(trifluoromethyl)-1,3-diazine
Density 1.879
Flash Point 53-54/5mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point -6 to -5
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-4-trifluoromethylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-4-trifluoromethylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart