APO497530926 Apollo Scientific Ltd

2,2-Difluoroethyl chloroformate

  • MDL number MFCD24141311
  • Cas Number : 111022-16-3
  • Molecular Weight : 144.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 72-74/150mm
Product Link https://store.apolloscientific.co.uk/product/22-difluoroethyl-chloroformate

Safety Information

signalword Danger
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluoroethyl-chloroformate
pictograms GHS02;GHS05;GHS06
hstatements H226;H311;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart