APO497530928 Apollo Scientific Ltd

2,2-Difluoroethyl 1H-imidazole-1-carboxylate

  • MDL number MFCD28334494
  • Cas Number : 1980034-77-2
  • Molecular Weight : 176.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-difluoroethyl 1H-imidazole-1-carboxylate
Inchi Code 1S/C6H6F2N2O2/c7-5(8)3-12-6(11)10-2-1-9-4-10/h1-2,4-5H,3H2
Inchi Key LDSBPOFJBICNEO-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22-difluoroethyl-1h-imidazole-1-carboxylate

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/22-difluoroethyl-1h-imidazole-1-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart