APO497692902 Apollo Scientific Ltd

1,5-Dimethyl-1H-pyrazole-4-boronic acid

  • MDL number MFCD13195678
  • Cas Number : 1204333-57-2
  • Molecular Weight : 139.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-1,5-dimethyl-1H-pyrazole
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/15-dimethyl-1h-pyrazole-4-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/15-dimethyl-1h-pyrazole-4-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart