APO497692907 Apollo Scientific Ltd

2,4-Dihydroxybenzonitrile

  • MDL number MFCD02261931
  • Cas Number : 64419-24-5
  • Molecular Weight : 135.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179
Synonym 4-Cyanobenzene-1,3-diol
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dihydroxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dihydroxybenzonitrile
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart