APO497692908 Apollo Scientific Ltd

(8S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride

  • MDL number MFCD17926204
  • Cas Number : 865303-57-7
  • Molecular Weight : 221.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-THQ-amine dihydrochloride
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8s-5678-tetrahydroquinolin-8-amine-dihydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8s-5678-tetrahydroquinolin-8-amine-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart