APO497692927 Apollo Scientific Ltd

4-(Difluoromethyl)aniline hydrochloride

  • MDL number MFCD28145075
  • Cas Number : 1980063-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Aminobenzal fluoride hydrochloride, alpha,alpha-Difluoro-p-toluidine hydrochloride
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethylaniline-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethylaniline-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart