APO498106247 Apollo Scientific Ltd

4-(Hydroxymethyl)benzaldehyde

  • MDL number MFCD11870191
  • Cas Number : 52010-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Synonym 4-Formylbenzyl alcohol, (4-Formylphenyl)methanol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-hydroxymethylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxymethylbenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart