APO498106295 Apollo Scientific Ltd

3-Cyclopropylphenol

  • MDL number MFCD06802406
  • Cas Number : 28857-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Hydroxyphenyl)cyclopropane
Purity 97
Flash Point >70
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 80-82/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50946

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50946
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart