APO498106322 Apollo Scientific Ltd

1,3-Bis(pentafluorosulphanyl)-5-bromobenzene

  • MDL number MFCD18071067
  • Cas Number : 432028-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-67
Synonym 1,3-Bis(pentafluorothio)-5-bromobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/13-bispentafluorosulphanyl-5-bromobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-bispentafluorosulphanyl-5-bromobenzene
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart