APO498106329 Apollo Scientific Ltd

2,6-Dichloro-4-fluorobenzaldehyde

  • MDL number MFCD20526366
  • Cas Number : 1182709-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-fluorobenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart