APO498106334 Apollo Scientific Ltd

2-Nitrobenzal fluoride

  • MDL number MFCD23382510
  • Cas Number : 64747-65-5
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Difluoromethyl)-2-nitrobenzene, 2-(Difluoromethyl)nitrobenzene, alpha,alpha-Difluoro-2-nitrotoluene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-nitrobenzal-fluoride

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-nitrobenzal-fluoride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart