APO513030284 Apollo Scientific Ltd

6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol

  • MDL number MFCD28740790
  • Cas Number : 1805580-01-1
  • Molecular Weight : 275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-2-fluoro-3-(trifluoromethoxy)phenol
Inchi Code 1S/C7H3BrF4O2/c8-3-1-2-4(5(9)6(3)13)14-7(10,11)12/h1-2,13H
Inchi Key YSYNWSMUZYYJRT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-2-fluoro-3-trifluoromethoxyphenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-2-fluoro-3-trifluoromethoxyphenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart