APO514206664 Apollo Scientific Ltd

4'-Hydroxy-2'-(trifluoromethyl)acetophenone

  • MDL number MFCD01091004
  • Cas Number : 220227-53-2
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[4-hydroxy-2-(trifluoromethyl)phenyl]ethanone
Inchi Code 1S/C9H7F3O2/c1-5(13)7-3-2-6(14)4-8(7)9(10,11)12/h2-4,14H,1H3
Inchi Key GKBDTFVRRWGDQM-UHFFFAOYSA-N

Description

Synonym 1-[4-Hydroxy-2-(trifluoromethyl)phenyl]ethanone
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylacetophenone
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart