APO514206665 Apollo Scientific Ltd

3-Bromo-2,4-difluorobenzonitrile

  • MDL number MFCD09261256
  • Cas Number : 935534-48-8
  • Molecular Weight : 218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-24-difluorobenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-24-difluorobenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart