APO514206667 Apollo Scientific Ltd

4-(Difluoromethoxy)-2-fluorobenzaldehyde

  • MDL number MFCD11847162
  • Cas Number : 1214358-05-0
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxy-2-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxy-2-fluorobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart