APO514505842 Apollo Scientific Ltd

3-Fluoro-4-nitropyridine-N-oxide

  • MDL number MFCD00234027
  • Cas Number : 769-54-0
  • Molecular Weight : 158.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-nitropyridine 1-oxide
Inchi Code 1S/C5H3FN2O3/c6-4-3-7(9)2-1-5(4)8(10)11/h1-3H
Inchi Key QHWIGULJOZAPAQ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-nitropyridine-n-oxide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-nitropyridine-n-oxide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart