APO514505874 Apollo Scientific Ltd

4-[(Difluoromethyl)sulphonyl]bromobenzene

  • MDL number MFCD13188442
  • Cas Number : 51679-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethylsulphonylbromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethylsulphonylbromobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart