APO514508707 Apollo Scientific Ltd

Disodium naphthalene-2,7-disulphonate

  • MDL number MFCD00066365
  • Cas Number : 1655-35-2
  • Molecular Weight : 332.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Naphthalene-2,7-disulphonic acid, disodium salt
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/disodium-naphthalene-27-disulphonate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/disodium-naphthalene-27-disulphonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart