APO514508710 Apollo Scientific Ltd

4-Bromo-3-iodonitrobenzene

  • MDL number MFCD17012734
  • Cas Number : 63037-63-8
  • Molecular Weight : 327.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-99
Synonym 1-Bromo-2-iodo-4-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-iodonitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-iodonitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart