APO516024148 Apollo Scientific Ltd

2-(2,5-Dichlorophenyl)-1,4-benzoquinone

  • MDL number MFCD00448731
  • Cas Number : 79756-69-7
  • Molecular Weight : 253.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-117
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-25-dichlorophenyl-14-benzoquinone

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-25-dichlorophenyl-14-benzoquinone
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart