APO517178358 Apollo Scientific Ltd

3,4-dimethylbenzene-1,2-diamine

  • MDL number MFCD00051709
  • Cas Number : 41927-01-9
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-90
Iupac Name 3,4-dimethyl-1,2-benzenediamine
Inchi Code 1S/C8H12N2/c1-5-3-4-7(9)8(10)6(5)2/h3-4H,9-10H2,1-2H3
Inchi Key MHQULXYNBKWNDF-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-90
Product Link https://store.apolloscientific.co.uk/product/34-dimethylbenzene-12-diamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-dimethylbenzene-12-diamine
pictograms GHS07
hstatements H302+H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart