APO517178365 Apollo Scientific Ltd

Silver bromide

  • MDL number MFCD00003398
  • Cas Number : 7785-23-1
  • Molecular Weight : 187.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 432
Iupac Name bromosilver
Inchi Code 1S/Ag.BrH/h;1H/q+1;/p-1
Inchi Key ADZWSOLPGZMUMY-UHFFFAOYSA-M

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 432
Product Link https://store.apolloscientific.co.uk/product/silver-bromide

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/silver-bromide
pictograms GHS09
hstatements H400;H410
Availability details for
Quantity
  • Availability
  • Add to Cart