APO517178385 Apollo Scientific Ltd

Ethyl 4-bromo-3-fluorobenzoate

  • MDL number MFCD09751983
  • Cas Number : 1130165-74-0
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-bromo-3-fluorobenzoate
Inchi Code 1S/C9H8BrFO2/c1-2-13-9(12)6-3-4-7(10)8(11)5-6/h3-5H,2H2,1H3
Inchi Key DDINDBNRZWUYBR-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-bromo-3-fluorobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-bromo-3-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart