APO517178387 Apollo Scientific Ltd

2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene

  • MDL number MFCD20526760
  • Cas Number : 1321963-74-9
  • Molecular Weight : 306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-1-iodo-4-(trifluoromethoxy)benzene
Inchi Code 1S/C7H3F4IO/c8-5-3-4(1-2-6(5)12)13-7(9,10)11/h1-3H
Inchi Key MAFILVJNIQSQSP-UHFFFAOYSA-N

Description

Synonym 3-Fluoro-4-iodophenyl trifluoromethyl ether
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-1-iodo-4-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-1-iodo-4-trifluoromethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart